HPLC Analysis of Ifosfamide Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Ifosfamide Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 C
mobile phase [A] water; [B] THF; (90:10, A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 5 µl
detector UV, 205 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antineoplastics, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC