Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Immunoglobulin G (IgG) on Zenix® SEC-150 versus Zenix® SEC-300, Effect of Pore Size on Resolution
HPLC Analysis of Immunoglobulin G (IgG) on Zenix® SEC-150 versus Zenix® SEC-300, Effect of Pore Size on Resolution
Conditions
column Zenix SEC-150, 30 cm x 4.6 mm I.D., 3 µm, 150 Angstrom particles (Z777016)
column Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 µm, 300 Angstrom particles (Z777028)
mobile phase 150 mM sodium phosphate, pH 7.0
flow rate 1 ml/min
sample 1 mg/mL, IgG
injection 10 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies