Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Immunoglobulins (IgM) on Q Sepharose® Fast Flow
HPLC Analysis of Immunoglobulins (IgM) on Q Sepharose® Fast Flow
Conditions
column 30 cm x 50 mm I.D. (Q1126-100ML)
media Q Sepharose Fast Flow (Q1126-100ML)
mobile phase [A] 0.02 M monosodium phosphate, pH 6.5; [B] A + 0.5 M sodium chloride, pH 6.5
flow rate 25 cm/hr
injection cell culture supernatant
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Sepharose is a registered trademark of GE Healthcare