Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Indapamide Enantiomers on Astec® Cellulose DMP, LC/MS
HPLC Analysis of Indapamide Enantiomers on Astec® Cellulose DMP, LC/MS
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 25 C
mobile phase [A] methanol; [B] ammonium formate; (100:0.1, A:B)
flow rate 0.5 ml/min
sample 2 mg/mL in mobile phase
injection 2 µL/h
detector UV, 230 nm
Description
Categories Analytical Chromatography, Diuretics
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC