HPLC Analysis of Inhibin α-Subunit Fragment 1-32 on Discovery® HS C18
HPLC Analysis of Inhibin α-Subunit Fragment 1-32 on Discovery® HS C18
Conditions
column Discovery HS C18, 10 cm x 2.1 mm ID, 5 mum particles (568501-U)
column temp. 22 °C
mobile phase [A] 20 mM formic acid in water; [B] (20 mM formic acid in water):(20 mM formic acid in acetonitrile), 50:50
flow rate 0.2 mL/min
sample 20 mg/L in 0.1 % formic acid
injection 2 µL
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC