Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Insulin (Species Variants) on Ascentis® Express Phenyl-Hexyl
HPLC Analysis of Insulin (Species Variants) on Ascentis® Express Phenyl-Hexyl
Conditions
column Ascentis Express Phenyl-Hexyl, 15 cm x 2.1 mm, 2.7 µm particles (53338-U)
column temp. 30 C
mobile phase [A] 70:30, water with 0.1% TFA:acetonitrile with 0.09% TFA; [B] 55:45, water with 0.1% TFA:acetonitrile with 0.09% TFA
gradient 0 to 100% B in 7.5 min
flow rate 0.3 ml/min
pressure 3960 psi (273 bar)
sample 200 µg/mL in 0.1% formic acid
injection 0.5 µl
detector UV, 215 nm
Description
Analysis Note These three insulins have more than 76% identical positions in a primary structure alignment. The human and porcine proteins are most similar with 85% identity. The Phenyl-Hexyl phase provides an alternate retention and selectivity to a C18 phase, particularly the C18 phase on a larger pore silica, Ascentis Express ES-C18
Categories Analytical Chromatography, Proteins, Peptides, Hormones
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)