HPLC Analysis of Isocyanate Derivatives on Ascentis® ES-Cyano
HPLC Analysis of Isocyanate Derivatives on Ascentis® ES-Cyano
Conditions
column Ascentis Cyano, 15 cm x 4.6 mm I.D., 5 µm particles (577306-U)
column temp. 35 °C
mobile phase [A] 0.1% ammonium acetate in water; [B] 0.1% ammonium acetate in acetonitrile; (70:30, A:B)
flow rate 1.0 mL/min
sample 20 µg/mL in 80:12:8, water: acetonitrile: dimethylsulfoxide
injection 5 µL
detector UV, 254 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Cyano for the efficient separation of isocyanate derivatives.
Categories Analytical Chromatography, Isocyanates
Featured Industry Industrial Hygiene