Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Isoflavones in Soy Milk on Ascentis® Express RP Amide (Milk Brand 1)
HPLC Analysis of Isoflavones in Soy Milk on Ascentis® Express RP Amide (Milk Brand 1)
Conditions
column Ascentis Express RP-Amide, 10 cm x 3 mm I.D., 2.7 µm particles (53918-U)
column temp. 30 C
mobile phase [A] 0.1% formic acid in water; [B] 0.1% formic acid in methanol
gradient held at 15% B for 1 min; 15 to 100% in 5 min; held at 100% B for 1 min; 100-15% B in 0.1 min; held at 15% B for 2.9 min
flow rate 0.8 ml/min
pressure 2900 psi (200 bar)
sample soy milk, methanol extract (1. Measure 2 mL of each brand of soy milk (6 samples) 2. Add 8 mL of methanol and shake for 1 hour 3. Centrifuge tubes at 10000 rpm for 30 minutes 4. Transfer aliquot to autosampler vial)
injection 10 µl
detector UV, 260 nm
Description
Analysis Note This application demonstrates the separation of seven isoflavones on Ascentis Express RP Amide
Categories Analytical Chromatography, Food & Beverage Analysis, Hormones, Beverage Testing, Milk
Featured Industry Food and Beverages
Vitamins, Nutraceuticals, and Natural Products