Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ketoconazole Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
HPLC Analysis of Ketoconazole Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
Conditions
column CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 µm particles (24024AST)
column temp. 25 C
mobile phase [A] 0.1% formic acid; [B] acetonitrile; (70:30, A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 1 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Antibiotics, Antimicrobials, Antifungals, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC