HPLC Analysis of Ketoconazole Enantiomers on Astec® Cellulose DMP, No Additives
HPLC Analysis of Ketoconazole Enantiomers on Astec® Cellulose DMP, No Additives
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 25 °C
mobile phase [A] acetonitrile; (100, A)
flow rate 0.5 mL/min
sample 2 mg/mL in mobile phase
injection 2 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Antifungals
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC