HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R (MS Detection)
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R (MS Detection)
Conditions
column CHIROBIOTIC R, 15 cm x 2.1 mm, 5 µm particles (13019AST)
column temp. 35 °C
mobile phase [A] 20 mM ammonium acetate, pH 5.6 [B] methanol; (70:30, A:B)
flow rate 0.2 mL/min
detector ESI(-)
Description
Categories Analytical Chromatography, Forensics and Toxicology, Pharmaceutical Analysis, Chiral Applications, Clinical, anti-inflammatories, Pharmaceutical (small molecule), Clinical, Forensic & Toxicology, Anti-inflammatories, All Pharmaceutical Applications, Therapeutic Drug Monitoring
Featured Industry Clinical
Forensics and Toxicology
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC