HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R
Conditions
column CHIROBIOTIC R, 15 cm x 4.6 mm I.D., 5 µm particles (13023AST)
column temp. 25 C
mobile phase [A] 20 mM ammonium acetate, pH 5.6; [B] methanol; (50:50, A:B)
flow rate 0.6 ml/min
sample 1 mg/mL in methanol
injection 5 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Anti-inflammatories, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC