HPLC Analysis of L-Cysteine on SeQuant ZIC-HILIC
HPLC Analysis of L-Cysteine on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 100 x 4.6 mm, 5 µm, 200 Ã… (1.50453.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] ammonium acetate; (80:20, A:B)
flow rate 0.5 mL/min
sample 1 mg/mL of acetylcysteine in acetonitrile/water (70/30)
injection 1 µL
detector 220 nm
Description
Categories Analytical Chromatography, Amino Acids