HPLC Analysis of β-Lactoglobulin, Carboxymethylated, Tryptic Digest on Discovery® BIO Wide Pore C18
HPLC Analysis of β-Lactoglobulin, Carboxymethylated, Tryptic Digest on Discovery® BIO Wide Pore C18
Conditions
column Discovery BIO Wide Pore C18, 15 cm x 2.1 mm I.D., 3 µm particles (567202-U)
column temp. 35 C
mobile phase [A] water/0.1% TFA; [B] (water/0.1% TFA):(acetonitrile/0.1% TFA), 50:50
flow rate 0.20 ml/min
injection 5 µl
detector UV, 215 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC