HPLC Analysis of Lansoprazole (Degradation Profile) on Ascentis® Express C18
HPLC Analysis of Lansoprazole (Degradation Profile) on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6mm, 2.7 µm particles (53829-U)
column temp. ambient
mobile phase [A] water, [B] acetonitrile:0.5% triethylamine in water, pH=7.0 (80:20)
flow rate 0.8 ml/min
injection 15 µl
detector UV, 285 nm
Description
Categories Analytical Chromatography, Antiulcer Compounds