HPLC Analysis of Lansoprazole Enantiomers on Astec® CYCLOBOND® I 2000 DMP
HPLC Analysis of Lansoprazole Enantiomers on Astec® CYCLOBOND® I 2000 DMP
Conditions
column CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 µm particles (20724AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] methanol; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.4:0.1, A:B:C:D)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antiulcer Compounds, Pharmaceutical (small molecule), Antiulcer Drugs, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC