Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Large DNA Fragments Containing More Than 10kb on TSKgel® DNA-STAT
HPLC Analysis of Large DNA Fragments Containing More Than 10kb on TSKgel® DNA-STAT
Conditions
column TSKgel DNA-STAT, 10 cm x 4.6 mm I.D., 5 µm particles (821962)
mobile phase [A] 0.02 M Tris-HCl, pH 8.5; [B] 1.0 M NaCl, pH 8.5
gradient 75 to 100% B in 20 min
flow rate 0.5 ml/min
sample 1 kbase ladder
detector UV, 260 nm
Description
Categories Analytical Chromatography, Nucleic Acids
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.