Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Large RNAs on TSKgel® Phenyl-5PW
HPLC Analysis of Large RNAs on TSKgel® Phenyl-5PW
Conditions
column TSKgel(R) Phenyl-5PW, 7.5 cm x 7.5 mm I.D., 10 µm particles (807573)
mobile phase [A] 2 M ammonium sulfate, 0.1 M phosphate buffer, pH 7.0; [B] 0.1 M phosphate buffer, pH 7.0
gradient 0 to 100% B in 60 min
flow rate 0.5 ml/min
sample 16S and 23S rRNA from E. coli, 0.5 mg/mL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Nucleic Acids
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.