HPLC Analysis of Linear Oligosaccharides on Chromolith® NH2
HPLC Analysis of Linear Oligosaccharides on Chromolith® NH2
Conditions
column Chromolith NH2, 10 cm x 4.6 mm I.D., 2 µm particles (1.52028.0001)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] water; (65:35, A:B)
flow rate 1 mL/min
pressure 377 psi (26 bar)
sample maltose 21.6 mg/mL, maltotriose 19.5 mg/mL, maltotetraose 15.7 mg/mL, maltopentaose 16.2 mg/mL, maltoheptaose 10.6mg/mL in mobile phase
injection 2 µL
detector UV, 190 nm
Description
Categories Analytical Chromatography, Carbohydrates
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany