Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Lorazepam Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Lorazepam Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 23 C
mobile phase methanol
flow rate 2 ml/min
sample 2 mg/mL in methanol
injection 5 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Anxiolytics, Sedatives, All Pharmaceutical Applications, Anxiolytics/Sedatives
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC