HPLC Analysis of Lorazepam Enantiomers on Astec® (R,R) P-CAP™
HPLC Analysis of Lorazepam Enantiomers on Astec® (R,R) P-CAP™
Conditions
column Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 µm particles (31024AST)
column temp. 25 °C
mobile phase [A] ethanol; [B] heptane; (50:50, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Anxiolytics, Sedatives, Pharmaceutical (small molecule), All Pharmaceutical Applications, Anxiolytics/Sedatives
Featured Industry Pharmaceutical (small molecule)
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC