HPLC Analysis of Luciferin Enantiomers on CHIRALPAK® AGP
HPLC Analysis of Luciferin Enantiomers on CHIRALPAK® AGP
Conditions
column CHIRALPAK AGP, 10 cm x 4 mm I.D., 5 µm particles (58150AST)
column temp. 25 C
mobile phase 10 mM sodium phosphate, pH 6.0
flow rate 0.9 ml/min
sample luciferin
detector UV, 225 nm
Description
Categories Analytical Chromatography, Chiral Applications, Vitamins, Nutraceuticals & Natural Products
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIRALPAK is a registered trademark of Daicel Corp.