HPLC Analysis of Luciferin Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Luciferin Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. ambient
mobile phase [A] 20 mM ammonium nitrate, pH 5.5; [B] THF; (90:10, A:B)
flow rate 1 ml/min
sample 0.5 mg/mL in methanol
injection 6 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Life Science & Biopharma, Other Biochemical Applications, All Biochemical Applications
Featured Industry Chemicals and Industrial Polymers
Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC