Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Lysozyme, Myoglobin and Impurities on Discovery® BIO Wide Pore C5
HPLC Analysis of Lysozyme, Myoglobin and Impurities on Discovery® BIO Wide Pore C5
Conditions
column Discovery BIO Wide Pore C5, 15cm x 4.6mm I.D., 5mum particles (568422-U)
column temp. ambient
mobile phase [A] (75:25) 0.1% TFA in water:0.1% TFA in acetonitrile; [B] (25:75) 0.1% TFA in water:0.1% TFA in acetonitrile
gradient 0-100% B in 25 min
flow rate 1.0 ml/min
injection 12 µl in 0.1% TFA
detector UV, 220 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC