Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of MAb 016 on Proteomix® SCX NP5 versus Competitive SCX using a Salt Gradient
HPLC Analysis of MAb 016 on Proteomix® SCX NP5 versus Competitive SCX using a Salt Gradient
Conditions
column Proteomix SCX-NP5, 25 cm x 4.6 mm I.D. (PEEK hardware), 5 µm particles (Z777164)
mobile phase (A) 20 mM phosphate buffer, pH 7.5; (B) A + 1 M NaCl
gradient 0 to 6% B in 30 min
flow rate 0.8 ml/min
sample MAb 016 (11.8 mg/mL)
injection 4 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies