HPLC Analysis of mAb F on Zenix®-300 and Zenix®-C 300
HPLC Analysis of mAb F on Zenix®-300 and Zenix®-C 300
Conditions
column Zenix-C SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777092)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777033)
column temp. 25 °C
mobile phase 150 mM sodium phosphate, pH 7.0
flow rate 1.0 mL/min
pressure 1073 psi (74 bar)
sample 1.23 mg/mL mAb F in 10 mM sodium succinate, pH 5.0
injection 10 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies