HPLC Analysis of mAb on Antibodix® WCX-NP5, 25 cm Columns
HPLC Analysis of mAb on Antibodix® WCX-NP5, 25 cm Columns
Conditions
column Antibodix WCX-NP5, 25 cm x 4.6 mm I.D., 5 µm particles (Z777294)
column temp. ambient
mobile phase (A) 10 mM sodium phosphate buffer, pH 7.5; (B) A + 100 mM NaCl
gradient 15 to 55% B in 30 min
flow rate 0.8 mL/min
sample monoclonal antibody
injection 10 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Antibodix is a registered trademark of Sepax Technologies