HPLC Analysis of mAb-X22 on Antibodix® WCX-NP10, Impact of Initial Salt Concentration
HPLC Analysis of mAb-X22 on Antibodix® WCX-NP10, Impact of Initial Salt Concentration
Conditions
column Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 µm particles (Z777272)
column temp. 25 °C
mobile phase (A) 10 mM sodium phosphate buffer, pH 7.5; (B), A + 100 mM NaCl
gradient 15 to 65% B in 60 min
flow rate 0.8 mL/min
sample MAb-X22, 1.5 mg/mL
injection 10 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Antibodix is a registered trademark of Sepax Technologies