HPLC Analysis of mAb-X22 on Antibodix® WCX-NP10, Impact of Mobile Phase pH
HPLC Analysis of mAb-X22 on Antibodix® WCX-NP10, Impact of Mobile Phase pH
Conditions
column Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 µm particles (Z777272)
column temp. 25 C
mobile phase (A) 10 mM sodium phosphate buffer at 7.5 pH; (B), A + 100 mM NaCl at 7.0 pH
gradient 15 to 65% B in 60 min
flow rate 0.8 ml/min
sample MAb-X22, 1.5 mg/mL
injection 10 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Antibodix is a registered trademark of Sepax Technologies