HPLC Analysis of Mandelic Acid Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Mandelic Acid Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 25 °C
mobile phase 0.1 wt% ammonium formate in methanol
flow rate 1 mL/min
sample 2 mg/mL in methanol
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Chemicals and Industrial Polymers, Organic Acids
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC