HPLC Analysis of Metabolites of 7,12-Dimethylbenz[a]anthracene on SUPELCOSIL™ LC-18
HPLC Analysis of Metabolites of 7,12-Dimethylbenz[a]anthracene on SUPELCOSIL™ LC-18
Conditions
column SUPELCOSIL LC-18, 25 cm x 4.6 mm I.D., 5 µm particles (58298)
mobile phase [A] methanol:[B] water, (50:50, A:B), 10 min to 100:0 at 2.5% /min
flow rate 0.8 ml/min
sample rat liver, 9000 x g supernatant fraction from Aroclor-treated animals
detector fluorescence
Description
Categories Analytical Chromatography, Forensics and Toxicology, Clinical, Environmental Analysis, hydrocarbons, Hydrocarbons, Organics, Chromatograms, HPLC Chromatograms
Featured Industry Environmental
Forensics and Toxicology
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC