HPLC Analysis of Methadone Enantiomers and Methadone Metabolite EDDP on Astec® CYCLOBOND® I 2000 HP-RSP
HPLC Analysis of Methadone Enantiomers and Methadone Metabolite EDDP on Astec® CYCLOBOND® I 2000 HP-RSP
Conditions
column CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 µm particles (24024AST)
mobile phase [A] 0.1% acetic acid; [B] acetonitrile; (80:20, A:B)
flow rate 0.8 ml/min
detector UV, 292 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Chiral Applications, analgesics, Clinical, Forensic & Toxicology, Drugs of Abuse, drugs of abuse, Pharmaceutical (small molecule), Pain Management, Therapeutic Drug Monitoring, All Pharmaceutical Applications
Featured Industry Forensics and Toxicology
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC