HPLC Analysis of Methadone Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Methadone Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 20 °C
mobile phase [A] methanol; [B] 20 mM ammonium formate; (95:5, A:B)
flow rate 1 mL/min
sample methadone, 1 mg/mL in mobile phase
injection 2 µL
detector UV, 205 nm
Description
Categories Analytical Chromatography, Clinical, Forensics and Toxicology, Pharmaceutical Analysis, Chiral Applications, Clinical, Forensic & Toxicology, drugs of abuse, Pharmaceutical (small molecule), Drugs of Abuse, analgesics, Analgesics, All Pharmaceutical Applications, Therapeutic Drug Monitoring, Pain Management
Featured Industry Clinical
Forensics and Toxicology
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC