HPLC Analysis of Methcathinone and Ephedrine on Discovery® HS F5
HPLC Analysis of Methcathinone and Ephedrine on Discovery® HS F5
Conditions
column Discovery HS F5, 15 cm x 4.6 mm I.D., 5 µm particles (567516-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate (pH 6.98); [B] acetonitrile (30:70, A:B)
flow rate 2 mL/min
injection 5 µL
detector photodiode array
Description
Categories Analytical Chromatography, Forensics and Toxicology, Pharmaceutical Analysis, stiµLants, bronchodilators, Alkaloids, decongestants
Featured Industry Forensics and Toxicology
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC