HPLC Analysis of Methionine Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Methionine Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 25 °C
mobile phase [A] water; [B] acetonitrile; (25:75, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Native Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC