HPLC Analysis of Methorphan Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Methorphan Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 25 °C
mobile phase 0.05 wt% ammonium trifluoroacetic acid in methanol
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 1 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antitussives, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC