HPLC analysis of Methyl Paraben and Organic Acids on Ascentis® Express RP-Amide
HPLC analysis of Methyl Paraben and Organic Acids on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (50774-U)
column temp. 25 °C
mobile phase [A] 0.1% formic acid; [B] acetonitrile; (70:30, A:B)
flow rate 1.3 mL/min
pressure 1870 psi (129 bar)
sample [A] water; [B] acetonitrile; (50:50, A:B)
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Antifungals
Featured Industry Flavors and Fragrances
Food and Beverages