HPLC Analysis of Methyl Phenyl Sulfoxide Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Methyl Phenyl Sulfoxide Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 25 °C
mobile phase methanol
flow rate 0.48 mL/min
sample 1 mg/mL in mobile phase
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Chemicals, Chemicals and Industrial Polymers, Other Organic Compound Applications
Featured Industry Chemicals and Industrial Polymers
Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC