Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of α-Methyl-α-Phenylsuccinimide Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of α-Methyl-α-Phenylsuccinimide Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 23 C
mobile phase [A] hexane; [B] ethanol; (80:20, A:B)
flow rate 1 ml/min
sample 5 mg/mL in methanol
injection 2 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Chemicals and Industrial Polymers, Organic Bases, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC