HPLC Analysis of α-Methylbenzylamine Enantiomers (3,5-Dinitrobenzoyl Derivatives) on Astec® CHIROBIOTIC® V
HPLC Analysis of α-Methylbenzylamine Enantiomers (3,5-Dinitrobenzoyl Derivatives) on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 C
mobile phase [A] hexane; [B] isopropanol; (90:10, A:B)
flow rate 0.5 ml/min
sample 2 mg/mL in methanol
injection 1 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Chemicals and Industrial Polymers, Organic Bases, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC