HPLC Analysis of Methylphenidate Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Methylphenidate Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 °C
mobile phase [A] methanol; [B] 1% triethylamine, pH 4/1; (95:5, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 2 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Chiral Applications, Clinical, Forensic & Toxicology, AD/HD Drugs, psychostiµLant, Pharmaceutical (small molecule), ADHD drugs, Drugs of Abuse, All Pharmaceutical Applications, Therapeutic Drug Monitoring
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC