Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Methylphenobarbital (Mephobarbital) Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Methylphenobarbital (Mephobarbital) Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 C
mobile phase [A] ethanol; [B] hexane; (25:75, A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 10 µl
detector UV, 205 nm
Description
Analysis Note Methylphenobarbital (also known as mephobarbital and mephobarbitone) is a barbiturate derivative used primarily as an anticonvulsant, but also as a sedative and anxiolytic. It is a chiral compound and the enantiomers have been shown to exhibit different bioactivity, one narcotic, one convulsant, neither of which is seen in the racemate. Shown here is the rapid, efficient separation of methylphenobarbital enantiomers on an Astec CHIROBIOTIC V column. Highest grade HPLC solvents were used to supply low background interference and low particulate contamination for robust, trouble-free operation. A Sigma-Aldrich USP reference standard provided reliable identification and quantification.
Categories Analytical Chromatography, Pharmaceutical Analysis, Chiral Applications, Pharmaceutical (small molecule), Anxiolytics, Sedatives, Anticonvulsant/antiepiletic/antiseizure, Anxiolytics/Sedatives, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC