HPLC Analysis of Methylselenocysteine Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Methylselenocysteine Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 25 °C
mobile phase [A] water; [B] acetonitrile; (30:70, A:B)
flow rate 1 mL/min
sample 2 mg/mL in methanol
injection 5 µL
detector UV at 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Antineoplastics, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC