HPLC Analysis of Mianserin Enantiomers on Astec® Cellulose DMP (No Additives)
HPLC Analysis of Mianserin Enantiomers on Astec® Cellulose DMP (No Additives)
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 25 C
mobile phase [A] acetonitrile; (100, A)
flow rate 0.5 ml/min
sample 2 mg/mL in mobile phase
injection 2 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, Antidepressants
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC