HPLC Analysis of Milk Proteins on HiTrap® SP
HPLC Analysis of Milk Proteins on HiTrap® SP
Conditions
column HiTrap SP, 1 mL (54817)
mobile phase [A] 50 mM sodium acetate, pH 6.0; [B] 50 mM sodium acetate/1.0 M sodium chloride, pH 6.0
flow rate 1 ml/min
injection 0.5 mL filtered casein-precipitated human milk (buffer exchanged to mobile phase A)
detector UV, 280 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Proteins, Beverage Testing, Milk
Featured Industry Food and Beverages
Life Science and Biopharma
Legal Information HiTrap is a registered trademark of GE Healthcare