HPLC Analysis of Monoclonal Antibodies (mAb) with Charge Variance on Antibodix® WCX-NP10
HPLC Analysis of Monoclonal Antibodies (mAb) with Charge Variance on Antibodix® WCX-NP10
Conditions
column Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 µm particles (Z777272)
column temp. ambient
mobile phase (A) 10 mM sodium phosphate buffer, pH 7.5; (B) A + 100 mM NaCl
gradient 15 to 55% B in 30 min
flow rate 0.8 ml/min
pressure 840 psi (58 bar)
sample monoclonal antibody
injection 5 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Antibodix is a registered trademark of Sepax Technologies