HPLC Analysis of Monosaccharides Ribose, Xylitol, and Mannose on Chromolith® NH2
HPLC Analysis of Monosaccharides Ribose, Xylitol, and Mannose on Chromolith® NH2
Conditions
column Chromolith NH2, 10 cm x 4.6 mm I.D., 2 µm particles (1.52028.0001)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] water; (85:15, A:B)
flow rate 3 mL/min
pressure 47 bar (682 psi)
sample ribose 5.9 mg/mL, xylitol 8.1 mg/mL, mannose 7.6 mg/mL in mobile phase
injection 5 µL
detector UV, 190 nm
Description
Categories Analytical Chromatography, Carbohydrates
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany