HPLC Analysis of Morphine and its 3- and 6-Glucuronide Metabolites on SeQuant ZIC-HILIC
HPLC Analysis of Morphine and its 3- and 6-Glucuronide Metabolites on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 50 x 4.6 mm, 5 µm, 200 Ã… (1.50451.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate 0.5 mL/min
pressure 763 psi (5.3 MPa)
injection 5 µL
detector 212 nm
Description
Featured Industry Clinical