HPLC Analysis of Mouse Ascites Fluid on TSKgel® BioAssist® Q vs. Competitive Q Column
HPLC Analysis of Mouse Ascites Fluid on TSKgel® BioAssist® Q vs. Competitive Q Column
Conditions
column TSKgel BioAssist Q, 5 cm x 4.6 mm I.D., 10 µm particles (819685)
column temp. 25 °C
gradient 15 min linear gradient of NaCl from 0 to 1 M in 20 mM Tris-HCl buffer, pH 8.0
flow rate 1 mL/min
sample mouse ascites fluid (3-fold dilution in 20 mM Tris-HCl buffer, pH 8.0)
injection 5 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information BioAssist is a registered trademark of Tosoh Corporation, TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.