HPLC Analysis of mPEG and mPEG-Peptide (Methoxy-PEG-Maleimide) on Zenix® and Zenix®-C SEC
HPLC Analysis of mPEG and mPEG-Peptide (Methoxy-PEG-Maleimide) on Zenix® and Zenix®-C SEC
Conditions
column Zenix-C SEC-300, 30 cm x 7.8 mm, I.D., 3 µm particles (Z777092)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777033)
column temp. 25 °C
mobile phase 150 mM sodium phosphate buffer, pH 7.0
flow rate 1.0 mL/min
sample 20kD peptide mPEG-MAL (methoxy-PEG-Maleimide), 1 mg/mL
injection 20 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Zenix is a registered trademark of Sepax Technologies